Learn More
Eppendorf™ Adapters For Eppendorf™ Rotor A-4-81
Product Code. 11893422 Shop All Eppendorf Products
Click to view available options
Capacity:
1.5 to 2 mL
2.6 to 5 mL
Unit Size:
Pack of 2
Product Code. 11893422
Brand: Eppendorf™ 5810746000
Add flexibility to Eppendorf Centrifuges with these rotor adapters
  • Adapters
  • For Eppendorf™ Rotor A-4-81
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Flat
Capacity 2.6 to 5 mL
For Use With (Equipment) Rotor A-4-81 Eppendorf
Bore Size (Metric) 11 mm
No. of Places 80
No. per Pack 2 Pack