Learn More
Thermo Scientific™ Adapters for Oval Single Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Product Code. p-6545041 Shop All Thermo Scientific Products
Click to view available options
Capacity:
6/8 x 250 mL Platelet-rich Plasma or Buffy Coat Bag
6/8 x 450 mL Double or Triple Blood Bag
6/8 x 550 mL Quad or Quint Blood Bag
Unit Size:
Each
Pack of 2
Product Code. 16679312
Brand: Thermo Scientific™ 75003838
Choice of 4 adapters to accommodate a range of blood bags in oval single blood bag buckets for Thermo Scientific™ HAEMAFlex™ 6/8 rotors.
Thermo Scientific™ blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.
Select adapters for 450–550mL Quad or Quint blood bags, 400–450mL Double or Triple blood bags, 250mL Platelet-rich Plasma or Buffy Coat bags or 400–450mL Single or Double blood bags.
TRUSTED_SUSTAINABILITY
Specifications
Type Blood Bag Adapter
Capacity 6/8 x 450 mL Double or Triple Blood Bag
For Use With (Equipment) Oval single blood bag buckets
No. per Pack Set of 2