missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAT2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82557-0.1ML
This item is not returnable.
View return policy
Description
ADAT2 Polyclonal specifically detects ADAT2 in Human samples. It is validated for Western Blot.
Specifications
| ADAT2 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 134637 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| adenosine deaminase, tRNA-specific 2, TAD2 homolog (S. cerevisiae), DEADC1, deaminase domain containing 1, Deaminase domain-containing protein 1, dJ20N2, dJ20N2.1, DKFZp686L1118, EC 3.5.4, EC 3.5.4.-, FLJ44213, TAD2, tRNA-specific adenosine deaminase 2, tRNA-specific adenosine deaminase 2 homolog, tRNA-specific adenosine-34 deaminase subunit ADAT2 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human ADAT2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction