missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADCK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545434
This item is not returnable.
View return policy
Description
Peptide sequence: LGNGRKPPEN LADQSFLERL LLPKADLVGS NAGVSRAQVP GHQPEATNLI Sequence homology: Human: 100%.
The function of this protein is not yet clear. It is not known if it has protein kinase activity and what type of substrate it would phosphorylate (Ser, Thr or Tyr).
Specifications
| ADCK2 | |
| Polyclonal | |
| Unconjugated | |
| ADCK2 | |
| AA408112; AARF; aarF domain containing kinase 2; Adck2; BB006057; MGC20727; putative ubiquinone biosynthesis protein AarF; uncharacterized aarF domain-containing protein kinase 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 90956 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7Z695 | |
| ADCK2 | |
| Synthetic peptide directed towards the middle region of human ADCK2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction