Learn More
Invitrogen™ ADCY1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597241
Description
Immunogen sequence: NIETFFIVPS HRRKIFPGLI LSDIKPAKRM KFKTVCYLLV QLMHCRKMFK AEIPFSNVMT CEDDDKRRAL RTASEKLRNR SSFSTNVVYT TPGTRVNRYI SRLLEARQTE LEMADLNFFT LKYKHVEREQ KYHQLQDEYF T; Positive Samples: Mouse brain, Mouse kidney; Cellular Location: Cell membrane, Cytoplasm, Membrane, Membrane raft, Multi-pass membrane protein.
ADCY1 belongs to the adenylyl cyclase class-4/guanylyl cyclase family. It is a calmodulin-sensitive adenylyl cyclase. ADCY1 may be involved in regulatory processes in the central nervous system. It may play a role in memory acquisition and learning.
Specifications
| ADCY1 | |
| Polyclonal | |
| Unconjugated | |
| ADCY1 | |
| 3',5'-cyclic AMP synthetase; Ac1; ADCY1; adenyl cyclase; adenylate cyclase 1; adenylate cyclase 1 (brain); adenylate cyclase type 1; Adenylate cyclase type I; adenylyl cyclase 1; ATP pyrophosphate-lyase 1; barrelless; brl; Ca(2+)/calmodulin-activated adenylyl cyclase; D11Bwg1392e; DFNB44; I-AC; mKIAA4070 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 107, 432530 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.63 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88444, Q08828 | |
| ADCY1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 470-610 of human ADCY1 (NP_0669391). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.