Learn More
Memmert™ Additional Stainless Steel Grid for IF450, Reinforced Chamber Option
Product Code. 17506985 Shop All Memmert Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. 17506985
Brand: Memmert™ IF450B32190
Only null left
Add to basket
Add to basket
Additional grid features a sturdy stainless steel construction.
  • Reinforced
  • Stainless Steel
  • Electropoloished
  • Max. loading 60 kg
Specifications
Capacity (Metric) Shelf 60 kg
For Use With (Equipment) IF450 Incubator
Material Stainless Steel
Type Reinforced Grid