Learn More
Memmert™ Additional Stainless Steel Grid for UF260plus, Reinforced Chamber Option
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF260plus Oven
Unit Size:
Each
Description
- Reinforced
- Stainless Steel
- Electropoloished
- Max. loading 60 kg
Specifications
Specifications
| For Use With (Equipment) | UF260plus Oven |
| Type | Reinforced Grid |
| Material | Stainless Steel |
| Max. Shelf Load | 60 kg |