Learn More
Memmert™ Additional Stainless Steel Shelf for IF450plus, Reinforced Chamber Option
Product Code. 17507025 Shop All Memmert Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. 17507025
Brand: Memmert™ IF450plusB32191
Only null left
Add to basket
Add to basket
Additional shelf features a sturdy stainless steel construction.
  • Reinforced
  • Stainless Steel
  • Max. loading 60 kg
Specifications
Capacity (Metric) Shelf 60 kg
For Use With (Equipment) IF450plus Incubator
Material Stainless Steel
Type Reinforced Shelf