Learn More
Memmert™ Additional Stainless Steel Shelf for UF450plus, Reinforced Chamber Option
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF450plus Oven
Unit Size:
Each
Description
- Reinforced
- Stainless Steel
- Max. loading 60 kg
Specifications
Specifications
| For Use With (Equipment) | UF450plus Oven |
| Type | Reinforced Shelf |
| Material | Stainless Steel |
| Max. Shelf Load | 60 kg |