Learn More
Memmert™ Additional Stainless Steel Shelf for UF450plus, Reinforced Chamber Option
Product Code. 17549375 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF450plus Oven
Unit Size:
Each
Product Code. 17549375
Brand: Memmert™ UF450plusB32191
Only null left
Add to basket
Add to basket
Additional shelf features a sturdy stainless steel construction.
  • Reinforced
  • Stainless Steel
  • Max. loading 60 kg
Specifications
For Use With (Equipment) UF450plus Oven
Type Reinforced Shelf
Material Stainless Steel
Max. Shelf Load 60 kg