Learn More
Adelmidrol, MedChemExpress
Product Code. 16468566
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16468566
Brand: Medchem Express HYB1026
MedChemExpress Adelmidrol exerts important anti-inflammatory effects that are partly dependent on PPARγ. Adelmidrol reduces NF-κB translocation, and COX-2 expression.
Adelmidrol exerts important anti-inflammatory effects that are partly dependent on PPARγ. Adelmidrol reduces NF-κB translocation, and COX-2 expression.
Specifications
Chemical Name or Material Adelmidrol
Physical Form Solid
CAS 1675-66-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H26N2O4
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : ≥ 100 mg/mL (364.48 mM)
SMILES O=C(NCCO)CCCCCCCC(NCCO)=O
Molecular Weight (g/mol) 274.36
Formula Weight 274.36
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only