missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine A1R Antibody (SR1610), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Adenosine A1R |
|---|---|
| Clone | SR1610 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671575
|
Novus Biologicals
NBP3-21593-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613185
|
Novus Biologicals
NBP3-21593-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adenosine A1R Monoclonal antibody specifically detects Adenosine A1R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
| Adenosine A1R | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| adenosine A1 receptor, ADORA 1, RDC7adenosine receptor A1 | |
| A synthesized peptide derived from human Adenosine A1R (Uniprot #: P30542) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1610 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, GPCR, Growth and Development, Immunology, Innate Immunity, Neuronal Cell Markers, Signal Transduction, Vision | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 134 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title