missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine A2b R Antibody (2683F), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB104442D
This item is not returnable.
View return policy
Description
Adenosine A2b R Monoclonal antibody specifically detects Adenosine A2b R in Human samples. It is validated for Flow Cytometry
Specifications
| Adenosine A2b R | |
| Monoclonal | |
| DyLight 350 | |
| adenosine A2b receptor, adenosine receptor A2b, ADORA2 | |
| Human Adenosine A2b R synthetic peptide, Accession # NP_000667 | |
| 0.1 mL | |
| Immunology, Signal Transduction | |
| 136 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry | |
| 2683F | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction