missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenovirus 3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93578MFV450
This item is not returnable.
View return policy
Description
Adenovirus 3 Polyclonal antibody specifically detects Adenovirus 3 in Nonspecies specific, Adenovirus, Virus samples. It is validated for Western Blot
Specifications
| Adenovirus 3 | |
| Polyclonal | |
| Western Blot | |
| ad3, adenovirus, Adenovirus 3 | |
| Synthetic peptide within residues 250-270 of the fiber knob protein of Ad 3. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Non-species specific, Adenovirus, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction