missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenovirus Antibody (8C4), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-413AF350
This item is not returnable.
View return policy
Description
Adenovirus Monoclonal antibody specifically detects Adenovirus in Adenovirus, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunodiffusion, ELISA, Lateral Flow, Neutralization
Specifications
| Adenovirus | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Adenovirus, Virus | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunodiffusion, ELISA, Lateral Flow, Blocking Assay | |
| 8C4 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunodiffusion, Sandwich ELISA, Lateral Flow Assay, Block/Neutralize | |
| Adenovirus | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human adenovirus type 1 and canine adenovirus type 1. | |
| 0.2 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction