missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenovirus Hexon Antibody (3G0), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65051R
This item is not returnable.
View return policy
Description
Adenovirus Hexon Monoclonal specifically detects Adenovirus Hexon in Adenovirus, Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Adenovirus Hexon | |
| Monoclonal | |
| DyLight 550 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| Recognizes the hexon group antigen present in most adenovirus serotypes. NB100-65051 has known reactivity with at least 17 serotypes of Adenovirus including types 40 & 41. | |
| Store at 4°C in the dark. |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 3G0 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Hexon protein; Late protein 2; PII | |
| ATCC VR5 virus | |
| Primary | |
| Adenovirus, Virus | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction