missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Adenovirus Hexon Monoclonal Antibody (F12C)

Description
The MA1-7326 antibody reacts with Adenovirus hexon in viral and infected tissue samples. MA1-7326 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7326 antibody was raised against Adenovirus hexon protein.
Hexon protein is a major coat protein of adenoviruses. Adenoviruses capsids have three principal protein components: the hexon, the penton, and the fiber. Hexon consists of three subunits together forming two major components of different morphological symmetry. A triangular top with three towers of density is superimposed on a more bulky pseudo hexagonal base.
Specifications
Specifications
| Antigen | Adenovirus Hexon |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | F12C |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Adeno Virus; AdV; HADV; HAdVC_gp08 |
| Host Species | Mouse |
| Immunogen | Adenovirus hexon protein. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?