missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 3 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
€ 357.00 - € 571.00
Specifications
| Antigen | Adenylate Cyclase 3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:5000-1:10000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653799
|
Novus Biologicals
NBP3-05527-100ul |
100 μg |
€ 571.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619248
|
Novus Biologicals
NBP3-05527-50ul |
50 μg |
€ 357.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adenylate Cyclase 3 Polyclonal antibody specifically detects Adenylate Cyclase 3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Adenylate Cyclase 3 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| AC3EC 4.6.1.1, AC-III, adenylate cyclase 3, adenylate cyclase type 3, Adenylate cyclase type III, Adenylyl cyclase 3, adenylyl cyclase, type III, ATP pyrophosphate-lyase 3, EC 4.6.1, KIAA0511, olfactive type | |
| C-terminal peptide of rat Adenlyate Cyclase 3, PAAFPNGSSVTLPHQVVDNP with a Cys added to the N-terminus to allow coupling to KLH. | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Chicken | |
| Lipid and Metabolism, Neuroscience | |
| 50% PBS, 50% glycerol | |
| 109 | |
| IgY | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title