missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Adenylate Cyclase 3 Monoclonal Antibody (1A12)
GREENER_CHOICE

Product Code. 17805127
Change view
Click to view available options
Quantity:
100 μL
500 μL
Unit Size:
100µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17805127 100 μL 100µL
17825107 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17805127 Supplier Invitrogen™ Supplier No. MA547381

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adenylate Cyclase 3 Monoclonal Antibody for ICC/IF, Western Blot

This gene encodes adenylyl cyclase 3 which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This protein appears to be widely expressed in various human tissues and may be involved in a number of physiological and pathophysiological metabolic processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Cyclase 3
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1A12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene Adcy3
Gene Accession No. O60266, P21932, Q8VHH7
Gene Alias AC3; AC-III; ADCY3; adenylate cyclase 3; adenylate cyclase 8 (brain); adenylate cyclase type 3; Adenylate cyclase type III; adenylate cyclase, olfactive type; Adenylyl cyclase 3; adenylyl cyclase, type III; ATP pyrophosphate-lyase 3; KIAA0511; mKIAA0511; type III adenylyl cyclase
Gene Symbols Adcy3
Host Species Mouse
Immunogen C-terminal peptide of rat ACIII, PAAFPNGSSVTLPHQVVDNP with a Cys added to the N-terminus to allow coupling to KLH.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104111, 109, 64508
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.