missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 5 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06369MFV450
This item is not returnable.
View return policy
Description
Adenylate Cyclase 5 Polyclonal antibody specifically detects Adenylate Cyclase 5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Adenylate Cyclase 5 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| AC5, adenylate cyclase 5, adenylate cyclase type 5, Adenylate cyclase type V, Adenylyl cyclase 5, ATP pyrophosphate-lyase 5, EC 4.6.1.1 | |
| Produced in rabbits immunized with E. coli-derived Human Adenylate Cyclase 5 fragment. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 111 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction