missing translation for 'onlineSavingsMsg'
Learn More

Adenylate Cyclase 8 Antibody (ADCY8/7341), FITC, Novus Biologicals™

Product Code. 18648544 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18648544 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18648544 Supplier Novus Biologicals Supplier No. NBP324064F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adenylate Cyclase 8 Monoclonal antibody specifically detects Adenylate Cyclase 8 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Cyclase 8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ADCY8/7341
Conjugate FITC
Formulation PBS
Gene Alias AC8, ADCY3, adenylate cyclase 8 (brain), adenylate cyclase type 8, Adenylate cyclase type VIII, Adenylyl cyclase 8, adenylyl cyclase-8, brain, ATP pyrophosphate-lyase 8, Ca(2+)/calmodulin-activated adenylyl cyclase, EC 4.6.1.1, HBAC1
Host Species Mouse
Immunogen Recombinant fragment (around aa1-200) of human Adenylate Cyclase 8 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 114
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.