missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 8 Antibody (ADCY8/7573), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24065UV
This item is not returnable.
View return policy
Description
Adenylate Cyclase 8 Monoclonal antibody specifically detects Adenylate Cyclase 8 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Adenylate Cyclase 8 | |
| Monoclonal | |
| DyLight 350 | |
| AC8, ADCY3, adenylate cyclase 8 (brain), adenylate cyclase type 8, Adenylate cyclase type VIII, Adenylyl cyclase 8, adenylyl cyclase-8, brain, ATP pyrophosphate-lyase 8, Ca(2+)/calmodulin-activated adenylyl cyclase, EC 4.6.1.1, HBAC1 | |
| Recombinant fragment (around aa1-200) of human Adenylate Cyclase 8 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 114 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| ADCY8/7573 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction