missing translation for 'onlineSavingsMsg'
Learn More

Adenylate Kinase 1 Antibody [mFluor Violet 500 SE], Novus Biologicals Biologicals™

Product Code. 30499284 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30499284 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30499284 Supplier Novus Biologicals Supplier No. NBP335229MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Adenylate Kinase 1 Polyclonal antibody specifically detects Adenylate Kinase 1 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Kinase 1
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias adenylate kinase 1, adenylate kinase isoenzyme 1, AK 1, ATP-AMP transphosphorylase 1, EC 2.7.4, EC 2.7.4.3, Myokinase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-194 of human Adenylate Kinase 1 (NP_000467.1).,, Sequence:, MEEKLKKTKIIFVVGGPGSGKGTQCEKIVQKYGYTHLSTGDLLRSEVSSGSARGKKLSEIMEKGQLVPLETVLDMLRDAMVAKVNTSKGFLIDGYPREVQQGEEFERRIGQPTLLLYVDAGPETMTQRLLKRGETSGRVDDNEETIKKRLETYYKATEPVIAFYEKRGIVRKVNAEGSVDSVFSQVCTHLDALK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 203
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.