missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylosuccinate Lyase Antibody (OTI2D10), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70101F
This item is not returnable.
View return policy
Description
Adenylosuccinate Lyase Monoclonal specifically detects Adenylosuccinate Lyase in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| Adenylosuccinate Lyase | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| ADSL | |
| Full length recombinant protein of human Adenylosuccinate Lyase (NP_000017) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry | |
| OTI2D10 | |
| Flow Cytometry, Immunohistochemistry | |
| adenylosuccinase, adenylosuccinate lyase, AMPS, ASase, ASL, EC 4.3.2, EC 4.3.2.2 | |
| Mouse | |
| Affinity Purified | |
| Core ESC Like Genes, Stem Cell Markers | |
| 158.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction