Learn More
3M™ Adflo™ Odor Filter
3M™ Adflo™ Odor Filter removes unpleasant smells and should be replaced any time you smell unpleasant odors.
Brand: 3M™ 7000028074
55.10 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 3.20000kg
Description
- Odor filter is not required if gas filter is used
- Snap-fastens between the blower unit and the particle filter
Specifications
Black | |
Odor Filter |