missing translation for 'onlineSavingsMsg'
Learn More

ADFP Mouse anti-Human, Clone: ADFP/1493, Abnova™

Product Code. 16033056
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16033056 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16033056 Supplier Abnova Supplier No. MAB14742.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human ADFP.

  • High specificity
  • High reactivity

Specifications

Antigen ADFP
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone ADFP/1493
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human ADFP.
Dilution ELISA Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS.
Gene Accession No. Q99541
Gene Alias ADRP/MGC10598
Gene Symbols ADFP
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 249-376 of human ADFP.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b λ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.