missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADFP Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA525042

Product Code. 13292659

  • € 578.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ADFP Polyclonal Antibody for Western Blot, IHC (P), Flow

Milk lipid globules from humans, cows and rats contain a protein identified as adipocyte differentiation-related protein (ADFP). It is associated with the globule surface membrane material. This protein, previously believed to be specific to adipocytes, is a major constituent of the globule surface and is present in a detergent-insoluble complex that contains butyrophilin and xanthine oxidase. ADFP (Adipophilin) is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells. In tissues, however, expression of adipophilin is restricted to specific cell types, such as lactating mammary epithelial cells, adrenal cortex cells, Sertoli and Leydig cells of the male reproductive system, and steatosis or fatty change hepatocytes in alcoholic liver cirrhosis. ADFP may be a possible new marker for the identification of specialized differentiated cells containing lipid droplets and for diseases associated with fat-accumulating cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ADFP
Polyclonal
Unconjugated
Plin2
AA407157; Adfp; Adipophilin; adipose differentiation related protein; adipose differentiation-related protein; ADPH; Adrp; ADRP antibody; MGC10598; perilipin 2; perilipin-2; Plin2
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
123
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q99541
Plin2
KLH conjugated synthetic peptide between 186-214 amino acids from the central region of human ADFP.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.