missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADGB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546236
This item is not returnable.
View return policy
Description
Peptide sequence: SESGGVSSPG KEEREQSTRK ENIQTGPRTR SPTILETSPR LIRKALEFMD Sequence homology: Cow: 80%; Human: 100%; Pig: 93%.
The function of this protein remains unknown.
Specifications
| ADGB | |
| Polyclonal | |
| Unconjugated | |
| ADGB | |
| ADGB; Androglobin; C6orf103; C9H6orf103; Calpain-7-like protein; CAPN16; CAPN7L | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79747 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N7X0 | |
| ADGB | |
| Synthetic peptide directed towards the C-terminal of human ADGB (aa 1490-1539). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction