missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADH5 Monoclonal Antibody (OTI4H10), TrueMAB™, OriGene
Description
This gene encodes a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. The encoded protein forms a homodimer. It has virtually no activity for ethanol oxidation, but exhibits high activity for oxidation of long-chain primary alcohols and for oxidation of S-hydroxymethyl-glutathione, a spontaneous adduct between formaldehyde and glutathione. This enzyme is an important component of cellular metabolism for the elimination of formaldehyde, a potent irritant and sensitizing agent that causes lacrymation, rhinitis, pharyngitis, and contact dermatitis. The human genome contains several non-transcribed pseudogenes related to this gene.
Specifications
Specifications
| Antigen | ADH5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI4H10 |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | ADH5 |
| Gene Accession No. | P11766 |
| Gene Alias | ADH-3, ADHX, FALDH, FDH, GSH-FDH, GSNOR, HEL-S-60p |
| Gene Symbols | ADH5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?