missing translation for 'onlineSavingsMsg'
Learn More

ADH5 Monoclonal Antibody (OTI4H10), TrueMAB™, OriGene

Product Code. 17531754
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17531754 30 μL 30µL
17511754 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17531754 Supplier Origene Technologies Supplier No. TA809479S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADH5 Monoclonal antibody specifically detects ADH5 in Human samples. It is validated for Western Blot

Specifications

Antigen ADH5
Applications Western Blot
Classification Monoclonal
Clone OTI4H10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ADH5
Gene Accession No. P11766
Gene Alias ADH1C; Adh2; Adh-2; ADH3; ADH-3; ADH5; Adh-5; ADH-B2; ADHX; alcohol dehydrogenase (class III), chi polypeptide; alcohol dehydrogenase 1C (class I), gamma polypeptide; alcohol dehydrogenase 2; Alcohol dehydrogenase 5; alcohol dehydrogenase 5 (class III), chi polypeptide; alcohol dehydrogenase B2; Alcohol dehydrogenase class chi chain; alcohol dehydrogenase class-3; Alcohol dehydrogenase class-III; class III alcohol dehydrogenase; class III alcohol dehydrogenase, chi subunit; epididymis secretory sperm binding protein Li 60p; FALDH; FDH; formaldehyde dehydrogenase; glutathione-dependent formaldehyde dehydrogenase; GSH-FDH; GSNOR; HEL-S-60p; S-(hydroxymethyl)glutathione dehydrogenase; S-nitrosoglutathione reductase
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-266 of human ADH5 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.