missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADH5 Mouse anti-Human, Clone: OTI4H3, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809455
This item is not returnable.
View return policy
Description
This gene encodes a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. The encoded protein forms a homodimer. It has virtually no activity for ethanol oxidation, but exhibits high activity for oxidation of long-chain primary alcohols and for oxidation of S-hydroxymethyl-glutathione, a spontaneous adduct between formaldehyde and glutathione. This enzyme is an important component of cellular metabolism for the elimination of formaldehyde, a potent irritant and sensitizing agent that causes lacrymation, rhinitis, pharyngitis, and contact dermatitis. The human genome contains several non-transcribed pseudogenes related to this gene.Specifications
| ADH5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11766 | |
| ADH5 | |
| Human recombit protein fragment corresponding to amino acids 1-266 of human ADH5 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4H3 | |
| Unconjugated | |
| ADH5 | |
| ADH-3, ADHX, FALDH, FDH, GSH-FDH, GSNOR, HEL-S-60p | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 128 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction