missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADH6 Polyclonal Antibody

Description
Immunogen sequence: AKEVRIKVVA TGLCGTEMKV LGSKHLDLLY PTILGHEG Highest antigen sequence indentity to the following orthologs: Rat - 61%, Mouse - 50%.
This gene encodes class V alcohol dehydrogenase, which is a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. This gene is expressed in the stomach as well as in the liver, and it contains a glucocorticoid response element upstream of its 5' UTR, which is a steroid hormone receptor binding site. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Antigen | ADH6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ADH6 |
| Gene Accession No. | P28332 |
| Gene Alias | 1810009C16Rik; ADH-5; Adh5a; ADH6; Adh6a; Alcohol dehydrogenase 6; alcohol dehydrogenase 6 (class V); alcohol dehydrogenase 6A (class V); aldehyde reductase; class V alcohol dehydrogenase |
| Gene Symbols | ADH6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?