missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADH7 Monoclonal Antibody (OTI2D11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526001
This item is not returnable.
View return policy
Description
ADH7 Monoclonal Antibody for Western Blot, IHC (P), Flow
This gene encodes class IV alcohol dehydrogenase 7 mu or sigma subunit, which is a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. The enzyme encoded by this gene is inefficient in ethanol oxidation, but is the most active as a retinol dehydrogenase; thus it may participate in the synthesis of retinoic acid, a hormone important for cellular differentiation. The expression of this gene is much more abundant in stomach than liver, thus differing from the other known gene family members.
Specifications
| ADH7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P40394 | |
| ADH7 | |
| Full length human recombinant protein of ADH7 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D11 | |
| Unconjugated | |
| ADH7 | |
| Adh3; Adh-3; Adh-3e; Adh3-e; Adh-3t; Adh3-t; Adh4; Adh7; ADH-C2; Adt-1; AI325182; alcohol dehydrogenase 3 complex; alcohol dehydrogenase 3 temporal; alcohol dehydrogenase 4; Alcohol dehydrogenase 7; alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide; alcohol dehydrogenase class 4 mu/sigma chain; Alcohol dehydrogenase class IV mu/sigma chain; alcohol dehydrogenase VII; alcohol dehydrogenase-7; All-trans-retinol dehydrogenase [NAD(+)] ADH7; class IV alcohol dehydrogenase, mu or sigma subunit; class IV sigma-1 alcohol dehydrogenase; class IV sigmasigma alcohol dehydrogenase; gastric alcohol dehydrogenase; IV ADH; Omega-hydroxydecanoate dehydrogenase ADH7; ret; Retinol dehydrogenase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 131 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction