Learn More
BTX™ Adherent Cell Electrodes
Click to view available options
Includes:
Cell Electrode Only
Cell Electrode, 450204 Cable
Unit Size:
Each
Description
- Specially designed to electroporate cultured monolayers of cells on laminin-coated filters, or on 12 mm polycarbonate or polyester transwell filter units (0.4 mm pore size)
- Can directly electroporate adherent cells such as human corneal keratocytes, Caco-2, dental pulp cells, NC65 and BCEC, and MDCK cells
- Consists of two parallel plate electrodes, 7 x 19 mm with 5 mm gap spacing between the plates
- Each plate electrode has a 0.3 mm height insulated foot at both ends of the bottom
- Compatible with BTX™ Systems such as Gemini X2 and ECM™ 2001+
Specifications
Specifications
| For Use With (Application) | Mammalian Cell Transfections, Plant and Yeast Applications, and Direct Electroporation Onto Adherent Cells Either Growing on Transwell Filters or Coverslips, or Directly Onto Monolayers Without Disruption |
| For Use With (Equipment) | ECM™ 830, ECM™ 2001+, Legacy ECM™ 2001, Gemini X2 |
| Product Type | Electrode Kit |
| Voltage | 300 V |
| Description | Adherent Cell Electrode Kit, 5 mm gap, includes 45-0204 cable |
| Field Type | Homogeneous |
| Includes | Cell Electrode, 450204 Cable |
| Pulse Length Range | 1 μsec.to 35 msec. |
| Quantity | 1 Ea. |
| Size (Metric) Gap | 5 mm |
| Show More |