missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene encodes an enzyme that belongs to the acireductone dioxygenase family of metal-binding enzymes, which are involved in methionine salvage. This enzyme may regulate mRNA processing in the nucleus, and may carry out different functions depending on its localization. A pseudogene that is located on chromosome 20 has been defined for this gene.
Specifications
Specifications
| Antigen | ADI1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.23 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ADI1 |
| Gene Accession No. | Q562C9, Q99JT9, Q9BV57 |
| Gene Alias | Acireductone dioxygenase, acireductone dioxygenase 1, ADI1, APL1, ARD, HMFT1638, MTCBP 1, MTCBP1, SIP L, SIPL |
| Gene Symbols | ADI1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?