missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADIG Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143174
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000. Preliminary analysis showed no signal in Western Blot.
Specifications
| ADIG | |
| Polyclonal | |
| Unconjugated | |
| ADIG | |
| ADIG; adipogenesis associated; adipogenin; SMAF1; small adipocyte factor 1 (SMAF1) | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 149685 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q0VDE8 | |
| ADIG | |
| Peptide with sequence SQESKEDDSDLC (aa 41-52). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction