missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adiponectin/Acrp30 Antibody (ADPN/1370), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54319IR
This item is not returnable.
View return policy
Description
Adiponectin/Acrp30 Monoclonal antibody specifically detects Adiponectin/Acrp30 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Specifications
| Adiponectin/Acrp30 | |
| Monoclonal | |
| DyLight 755 | |
| ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein | |
| Recombinant human Adiponectin/Acrp30 protein fragment (around aa 145-226) (exact sequence is proprietary) (Uniprot: Q15848) | |
| 100 μg | |
| Diabetes Research, Lipid and Metabolism | |
| 9370 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF | |
| ADPN/1370 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction