missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adiponectin Monoclonal Antibody (OTI2E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804681S
This item is not returnable.
View return policy
Description
Adiponectin Monoclonal antibody specifically detects Adiponectin in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| Adiponectin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15848 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9370 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2E2 | |
| Unconjugated | |
| ADIPOQ | |
| 30 kDa adipocyte complement-related protein; 30kDa; Acdc; Acrp30; Ad; adipo; adipocyte complement related protein; adipocyte complement related protein of 30 kDa; adipocyte complement-related 30 kDa protein; adipocyte complement-related protein of 30 kDa; adipocyte, C1Q and collagen domain containing; adipocyte, C1q and collagen domain-containing protein; Adipocyte-specific protein AdipoQ; adiponectin; adiponectin, C1Q and collagen domain containing; Adipoq; Adipose most abundant gene transcript 1 protein; adipose specific collagen-like factor; ADIPQTL1; ADPIOQ; ADPN; Apm1; APM-1; APN; GBP28; Gelatin-binding protein; gelatin-binding protein 28 | |
| Human recombinant protein fragment corresponding to amino acids 19-244 of human ADIPOQ produced in SF9 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction