missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adiponectin Monoclonal Antibody (OTI2G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804592
This item is not returnable.
View return policy
Description
Adiponectin is a hormone exclusively produced in adipose tissue and secreted into plasma. It is involved in a number of metabolic processes, such as glucose regulation, and the metabolism of fat for energy production. Adiponectin is involved in the control of fat metabolism and insulin sensitivity.Specifications
| Adiponectin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15848 | |
| ADIPOQ | |
| Human recombit protein fragment corresponding to amino acids 19-244 of human ADIPOQ produced in SF9 cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G8 | |
| Unconjugated | |
| ADIPOQ | |
| ACDC, ACRP30, ADIPQTL1, ADPN, APM-1, APM1, GBP28 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9370 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction