missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEUROD4 Monoclonal Antibody (OTI1B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810658S
This item is not returnable.
View return policy
Description
NEUROD4 Monoclonal antibody specifically detects NEUROD4 in Human samples. It is validated for Western BlotSpecifications
| NEUROD4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00221 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4794 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1B7 | |
| Unconjugated | |
| Nfkbie | |
| AI846749; ath3; ATH-3; Atoh3; atonal homolog 3; atonal-like 3; atonal-like protein 3; bHLH domain; bHLHa4; Class A basic helix-loop-helix protein 4; helix-loop-helix protein mATH-3; mATH3; MATH-3; NeuroD4; neurod4 protein; neurogenic differentiation 4; neurogenic differentiation factor 4; neuronal basic helix-loop-helix transcription factor Ath3; neuronal differentiation 4; protein atonal homolog 3; zath3 | |
| Full length human recombinant protein of human NFKBIE produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction