Learn More
Adiponectin Mouse anti-Human, Clone: OTI2C9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF804555
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Adiponectin is a hormone exclusively produced in adipose tissue and secreted into plasma. It is involved in a number of metabolic processes, such as glucose regulation, and the metabolism of fat for energy production. Adiponectin is involved in the control of fat metabolism and insulin sensitivity.Specifications
| Adiponectin | |
| Monoclonal | |
| Unconjugated | |
| ADIPOQ | |
| ACDC, ACRP30, ADIPQTL1, ADPN, APM-1, APM1, GBP28 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9370 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q15848 | |
| ADIPOQ | |
| Human recombit protein fragment corresponding to amino acids 19-244 of human ADIPOQ produced in SF9 cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.