missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADK Antibody (4F8), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97934AF350
This item is not returnable.
View return policy
Description
ADK Monoclonal specifically detects ADK in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| ADK | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ADK | |
| Recombinant human ADK (22-362aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 4F8 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| adenosine kinase, AKAdenosine 5′-phosphotransferase, EC 2.7.1, EC 2.7.1.20 | |
| Mouse | |
| Protein G purified | |
| Cancer, Cell Cycle and Replication, Immunology, Lipid and Metabolism | |
| 132.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction