missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADK Antibody (4F8), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97934PCP
This item is not returnable.
View return policy
Description
ADK Monoclonal specifically detects ADK in Human samples. It is validated for Flow Cytometry.
Specifications
| ADK | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| ADK | |
| Recombinant human ADK (22-362aa) purified from E. coli | |
| 100 μL | |
| Lipid and Metabolism | |
| 132 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 4F8 | |
| Flow Cytometry | |
| adenosine kinase, AKAdenosine 5'-phosphotransferase, EC 2.7.1, EC 2.7.1.20 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human ADK mAb, clone 4F8, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human ADK protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction