missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADK Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32189H
This item is not returnable.
View return policy
Description
ADK Polyclonal antibody specifically detects ADK in Human, Mouse samples. It is validated for Western Blot
Specifications
| ADK | |
| Polyclonal | |
| Western Blot | |
| adenosine kinase, AKAdenosine 5'-phosphotransferase, EC 2.7.1, EC 2.7.1.20 | |
| The immunogen this antibody was made to, maps to a region between residue 312 to 362 of human Adenosine Kinase using the numbering given in entry NP_006712.2 (GeneID 132). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Immunology, Lipid and Metabolism | |
| 132 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction