missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADNP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599221
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ADNP.
Vasoactive intestinal peptide is a neuroprotective factor that has a stimulatory effect on the growth of some tumor cells and an inhibitory effect on others. This gene encodes a protein that is upregulated by vasoactive intestinal peptide and may be involved in its stimulatory effect on certain tumor cells. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor. This gene is also upregulated in normal proliferative tissues. Finally, the encoded protein may increase the viability of certain cell types through modulation of p53 activity. Alternatively spliced transcript variants encoding the same protein have been described.
Specifications
| ADNP | |
| Polyclonal | |
| Unconjugated | |
| Adnp | |
| AA589558; activity dependent neuroprotector homeobox; activity-dependent neuroprotective protein; activity-dependent neuroprotector; activity-dependent neuroprotector homeobox; activity-dependent neuroprotector homeobox protein; Adnp; ADNP homeobox 1; ADNP1; HVDAS; KIAA0784; mKIAA0784; MRD28; NAP peptide | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11538, 23394 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H2P0, Q9Z103 | |
| Adnp | |
| A synthesized peptide derived from human ADNP(Accession Q9H2P0), corresponding to amino acid residues I125-P175. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction