missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADNP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17127448 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17127448 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17127448 Supplier Invitrogen™ Supplier No. PA5116210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ADNP2 Polyclonal Antibody for Western Blot

ADNP2 gene ontology annotations related to this gene include cellular response to oxidative stress; cellular response to retinoic acid; negative regulation of cell death; neuron differentiation; positive regulation of cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADNP2
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ADNP2
Gene Accession No. Q6IQ32, Q8CHC8
Gene Alias 8430420L05Rik; Activity-dependent neuroprotector homeobox protein 2; ADNP homeobox 2; ADNP homeobox protein 2; ADNP homeobox protein 2; activity-dependent neuroprotector homeobox protein 2; ADNP homeobox protein 2-like protein; ADNP2; BC024969; Kiaa0863; mKIAA0863; RGD1308279; Zfp508; Zinc finger protein 508; Znf508
Gene Symbols ADNP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 702-731 amino acids from the Central region of human ADNP2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22850, 240442
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.