missing translation for 'onlineSavingsMsg'
Learn More

ADPRM Polyclonal Antibody
GREENER_CHOICE

Product Code. 18035581
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18035581 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18035581 Supplier Invitrogen Supplier No. PA5144971

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ADPRM Polyclonal Antibody

ADPRM Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen ADPRM
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADPRM
Gene Accession No. Q3LIE5
Gene Alias 2310004I24Rik; ADPRibase-Mn; ADP-ribose/CDP-alcohol diphosphatase, manganese dependent; ADP-ribose/CDP-alcohol diphosphatase, manganese-dependent; ADP-ribose/CDP-alcohol pyrophosphatase; Adprm; C17orf48; CDP-choline phosphohydrolase; liver manganese (II)-dependent ADP-ribose/CDP-alcohol pyrophosphatase; manganese-dependent ADP-ribose/CDP-alcohol diphosphatase; Manganese-dependent ADP-ribose/CDP-alcohol diphosphatase-like protein; MDS006; NBLA03831; RGD1309906
Gene Symbols ADPRM
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 2-82. Recombinant protein control fragment (Product #RP-110109).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56985
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.