missing translation for 'onlineSavingsMsg'
Learn More

ADPS, MedChemExpress

Product Code. 16401796
Encompass_Preferred
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 16401796

Brand: Medchem Express HY15904

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ADPS is a kind of new Trinder's reagents, which are high water-soluble aniline derivatives, widely used in diagnostic and biochemical tests.

ADPS is a kind of new Trinder's reagents, which are high water-soluble aniline derivatives, widely used in diagnostic and biochemical tests.

Specifications

Chemical Name or Material ADPS
CAS 82611-88-9
Color Light Gray
Purity Grade Notes Research
Molecular Formula C12H18NNaO4S
Quantity 500 mg
Synonym ESPAS
Solubility Information H2O : ≥ 100 mg/mL (338.60 mM)
SMILES O=S(CCCN(CC)C1=CC=CC(OC)=C1)(O[Na])=O
Molecular Weight (g/mol) 295.33
Formula Weight 295.33
Percent Purity 98.51%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.