Learn More
ADRB3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30713-1-AP-150UL
 View more versions of this product
Product Code. 18050326
  • € 410.00 / 150µL
Description
The ADRB3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ADRB3. This antibody recognizes human antigen. The ADRB3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ADRB3
1100 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
ADRB3, ADRB3R, adrenergic, beta 3 , receptor, B3AR, Beta 3 adrenergic receptor, Beta 3 adrenoceptor, Beta 3 adrenoreceptor, BETA3AR, FLJ99960
150 μL
155
Store at -20°C. Stable for one year after shipment.