Learn More
ADRBK1 Mouse, anti-Human, CoraLite Plus 488, Clone: 2F7A5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67067-100UL
 View more versions of this product
Product Code. 18035215
  • € 450.00 / 100µL
Description
The ADRBK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ADRBK1. This antibody recognizes human antigen. The ADRBK1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
ADRBK1
2F7A5
CoraLite Plus 488
Mouse
Primary
Human
155 kDa
IgG2a
Western Blot
1000 μg/mL
ADRBK1, BARK, BARK1, Beta ARK 1, BETA ARK1, FLJ16718, GRK2
100 μL
156
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.