missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Adrenomedullin Monoclonal Antibody (OTI8A1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526819
This item is not returnable.
View return policy
Description
Adrenomedullin Monoclonal Antibody for IHC (P)
Adrenomedullin (ADM), a vasodilator produced by most contractile cells, is characterized by persistent hypotensive activity. ADM is involved in the regulation of fluid and electrolyte homeostasis and in the maintenance of cardiovascular functioning. In hypertensive patients, the level of ADM in plasma is upregulated. Natriuresis is a common systemic manifestation of aneurysmal subarachnoid hemorrhage. ADM has strong natriuretic actions. ADM-induced natriuresis is caused by an increase in glomerular filtration rate and a decrease in distal tubular sodium reabsorption. ADM is present both in the periphery and brain, and can exert central effects such as decreasing food ingestion.
Specifications
| Adrenomedullin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35318 | |
| ADM | |
| Human recombinant protein fragment corresponding to amino acids 22-146 of ADM produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin) | |
| OTI8A1 | |
| Unconjugated | |
| ADM | |
| ADM; Adrenomedullin; AM; AMpreproadrenomedullin; Ap; H39316; PAMP; preproadrenomedullin; proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide; ProAM-N20; RATAP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 133 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction